0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Potassium chloride
Potassium chloride 1 mol/l (1 N) in aqueous solution
Potassium chloride 1 mol/l (1 N) in aqueous solution
R005
:
G-Biosciences
:
Potassium chloride 1 mol/l (1 N) in aqueous solution
R005
:
G-Biosciences
:
Pack type:
Glass bottle
Pk:
100 mL
Seq:
2440
:
:
74.55 g/mol
:
1.13 g/cm³ (20 °C)
:
Ambient
:
231-211-8
:
7447-40-7
:
KCl
:
MFCD00011360