Order Entry
Czech Republic
ContactUsLinkComponent
Copper, BAKER ANALYZED® ACS, J.T.Baker®
Copper, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 1736-01
Supplier:  Avantor
CAS Number:  
undefined
Copper, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 1736-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.90 %
  • Assay (Cu):
    ≥ 99.90 %
  • Insoluble in Dilute HNO₃:
    ≤ 0.02 %
  • Antimony and Tin (as Sn):
    ≤ 0.005 %
  • Iron (Fe):
    ≤ 0.005 %
  • Lead (Pb):
    ≤ 0.005 %
  • Manganese (Mn):
    ≤ 0.001 %
  • Phosphorus (P):
    ≤ 0.001 %
  • Silver (Ag):
    ≤ 0.002 %
  • Trace Impurities - Arsenic (As):
    ≤ 5.000 ppm

Specification Test Results

About this item

 :
 :63.55 g/mol
Merck Index:13,02545
Boiling Pt:2595 °C (1013 hPa)
Melting Pt:1083 °C
Density:8.94 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-159-6
CAS Number:7440-50-8
Formula:Cu
MDL Number:MFCD00010965