
N-(1-Naphthyl)ethylenediamine dihydrochloride ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1465-25-4
:
Formula:C₁₂H₁₆Cl₂N₂
MDL Number:MFCD00012556
:259.18 g/mol
Merck Index:14,6409
Melting Pt:191…199 °C
EINECS:215-981-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together