L(-)-Tryptophan cell culture reagent
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:73-22-3
:
Formula:C₁₁H₁₂N₂O₂
MDL Number:MFCD00064340
:204.23 g/mol
Merck Index:14,9797
Melting Pt:280…285 °C
Density:1.34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-795-6
Related Information
Frequently Bought Together