0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
D(+)-Sucrose
D-(+)-Sucrose 99%, Ultrapure
D-(+)-Sucrose 99%, Ultrapure
:
Thermo Fisher Scientific
D-(+)-Sucrose 99%, Ultrapure
:
Thermo Fisher Scientific
:
57-50-1
:
:
C₁₂H₂₂O₁₁
:
MFCD00006626
:
14,8881
:
185…187 °C
:
1.58
:
342.3 g/mol
:
Ambient
:
200-334-9
Documentation
SDS
msds_ALFAJ64270
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Beakers, Low Form
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
D-(+)-Glucose, anhydrous 97.5-102.0% USP, Ph. Eur.
: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
TLC and GLP plates, silica gel 60, unmodified layers, glass backed
: Merck
Polysorbate 20, Reagent Grade
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Tubing connectors, T-shaped
: KARTELL
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Sodium chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
VWR®, Transfer Pipettes, High Performance
: VWR Collection