Order Entry
Czech Republic
ContactUsLinkComponent
Iron(III) nitrate nonahydrate, BAKER ANALYZED® ACS, J.T.Baker®
Oxidizing
Irritant
Iron(III) nitrate nonahydrate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 2018-01
Supplier:  Avantor
CAS Number:  
undefined
Oxidizing
Irritant
Iron(III) nitrate nonahydrate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 2018-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    98.0 - 101.0 %
  • Assay (Fe(NO₃)₃9H₂O) (by iodometry):
    98.0 - 101.0 %
  • Insoluble Matter:
    ≤ 0.005 %
  • Chloride (Cl):
    ≤ 5 ppm
  • Sulfate (SO₄):
    ≤ 0.01 %
  • Calcium (Ca):
    ≤ 0.01 %
  • Magnesium (Mg):
    ≤ 0.005 %
  • Potassium (K):
    ≤ 0.005 %
  • Sodium (Na):
    ≤ 0.05 %
  • Manganese (Mn):
    ≤ 0.01 %
  • pH of 5% Solution at 25°C:
    1.5 - 2.5

Specification Test Results

About this item

 :
 :404 g/mol
Merck Index:13,04057
Boiling Pt:125 °C (1013 hPa)
Melting Pt:47 °C
Density:1.68 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-899-5
UN:1466
ADR:5.1,III
 :GHS03
 :GHS07
CAS Number:7782-61-8
Formula:Fe(NO₃)₃·9H₂O
MDL Number:MFCD00149708