Order Entry
Czech Republic
ContactUsLinkComponent
 
Toxic
Health
Environment
Cadmium(II) sulphate 8/3-hydrate 99.0-102.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 1243-04
Supplier:  Avantor
CAS Number:  
undefined
Toxic
Health
Environment
Cadmium(II) sulphate 8/3-hydrate 99.0-102.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 1243-04
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    125 g
  • Seq:
    3445

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    99.0 - 102.0 %
  • Assay (CdSO₄.8/3 H₂O) (by EDTA titrn):
    99.0 - 102.0 %
  • Insoluble Matter:
    ≤0.005 %
  • Chloride (Cl):
    ≤0.001 %
  • Nitrate and Nitrite (as NO₃):
    ≤0.003 %
  • Calcium (Ca):
    ≤0.005 %
  • Copper (Cu):
    ≤0.002 %
  • Trace Impurities - Iron (Fe):
    ≤0.001 %
  • Lead (Pb):
    ≤0.001 %
  • Potassium (K):
    ≤0.01 %
  • Sodium (Na):
    ≤0.02 %
  • Zinc (Zn):
    ≤0.05 %

Specification Test Results

About this item

 :
 :256.51 g/mol (anhy)
Merck Index:13,01627
Melting Pt:41 °C
Density:3.08 kg/l
EINECS:233-331-6
UN:2570
ADR:6.1,III
 :GHS06
 :GHS08
 :GHS09
CAS Number:7790-84-3
Formula:3CdSO₄·8H₂O
MDL Number:MFCD02253000