

Pyrogallol, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0289-04
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, wide mouth, black T/E cap
- Pk:125 g
- Seq:3445
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:—
- Melting Point:131.0 - 135.0 °C
- Residue after Ignition:≤0.005 %
- Chloride (Cl):≤0.001 %
- Sulfate (SO₄):≤0.005 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Meets Reagent Specifications for testing USP/NF monographs:—
Specification Test Results
About this item
:
:126.11 g/mol
Merck Index:13,08090
Boiling Pt:309 °C (1013 hPa)
Melting Pt:130…135 °C
Density:1.453 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-762-9
UN:2811
ADR:6.1,III
:GHS08
:GHS07
CAS Number:87-66-1
Formula:C₆H₆O₃
MDL Number:MFCD00002192