4'-Methoxy-3'-nitroacetophenone, Sigma-Aldrich®
Catalog # SIAL553654-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:195.18 g/mol
:M-Clarity™ quality level = MQ100
EINECS:228-476-7
CAS Number:6277-38-9
Formula:C₉H₉NO₄
MDL Number:MFCD00017023