
Cerium(III) carbonate hydrate ≥99.999% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:54451-25-1
:
Formula:C₃Ce₂O₉·H₂O
MDL Number:MFCD00149635
:478.274 g/mol
Merck Index:14,1996
Melting Pt:848 °C
EINECS:208-655-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together