4-Bromobenzylamine hydrochloride, Sigma-Aldrich®
Catalog # SIAL205214-5G
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:5 g
- Sekvence:3173
Specifications
About this item
:
:222.52 g/mol
:M-Clarity™ quality level = MQ100
EINECS:247-503-3
CAS Number:26177-44-6
Formula:C₇H₉BrClN
MDL Number:MFCD00012860