3-Bromobenzylamine hydrochloride, Sigma-Aldrich®
Catalog # SIAL316288-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:222.52 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:39959-54-1
Formula:C₇H₉BrClN
MDL Number:MFCD00012856