Order Entry
Czech Republic
ContactUsLinkComponent
Potassium chloride 0.075 mol/l (0.075 N) in aqueous solution, Sigma-Aldrich®
Potassium chloride 0.075 mol/l (0.075 N) in aqueous solution, Sigma-Aldrich®
Catalog # SIALP9327-100ML
Supplier:  Merck
CAS Number:  
Potassium chloride 0.075 mol/l (0.075 N) in aqueous solution, Sigma-Aldrich®
Catalog # SIALP9327-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Potassiumchloride
 :
 :74.55 g/mol
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360