
3-Chloro-4-(trifluoromethyl)phenylboronic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:847756-88-1
:
Formula:C₇H₅BClF₃O₂
MDL Number:MFCD03788417
:224.38 g/mol
Melting Pt:206…212 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together