D-(+)-Galactosamine hydrochloride ≥99%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:1772-03-8
:
Formula:C₆H₁₄ClNO₅
MDL Number:MFCD00135830
:215.64 g/mol
Melting Pt:182…185 °C (dec., lit.)
EINECS:217-198-1
Specification Test Results
- Identity Test :Passes
- Purity:≥99%
- FTIR:Conforms to Standard
- Optical Rotation (c=1, H2O, after 24 hrs.):+93 ± 5°
Specification Test Results
Frequently Bought Together