


Hydrogen chloride 3 mol/l in cyclopentyl methyl ether
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
:36.46 g/mol
Boiling Pt:106 °C
Density:0.895
Flash Pt:10 °C (50 °F)
EINECS:231-595-7
UN:2924
ADR:3,II
:GHS02
:GHS06
:GHS05
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together