

4,6-Dichloro-o-toluidine ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:30273-00-8
:
Formula:C₇H₇Cl₂N
MDL Number:MFCD00044074
:176.05 g/mol
Boiling Pt:82…84 °C (0.1 mmHg)
Melting Pt:43…45 °C
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together