Order Entry
Czech Republic
ContactUsLinkComponent
Acetic anhydride-[1,1'-¹³C₂,²H₆] (99% ¹³C , 98% D), Sigma-Aldrich®
Acetic anhydride-[1,1'-¹³C₂,²H₆] (99% ¹³C , 98% D), Sigma-Aldrich®
Katalogové číslo SIAL607452-100MG
Supplier:  Merck
CAS Number:  
Acetic anhydride-[1,1'-¹³C₂,²H₆] (99% ¹³C , 98% D), Sigma-Aldrich®
Katalogové číslo SIAL607452-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
CAS Number:285977-78-8
Formula:C₄H₆O₃
MDL Number:MFCD00190252