Order Entry
Czech Republic
ContactUsLinkComponent
Barium-2-ethylhexanoate (∼17.5% Ba) in 2-ethylhexanoic acid
Toxic
Health
Barium-2-ethylhexanoate (∼17.5% Ba) in 2-ethylhexanoic acid
Catalog # 44847.14
CAS Number:  
Toxic
Health
Barium-2-ethylhexanoate (∼17.5% Ba) in 2-ethylhexanoic acid
Catalog # 44847.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :423.76 g/mol
EINECS:219-535-8
 :GHS06
 :GHS08
Storage Temperature:Ambient
CAS Number:2457-01-4
Formula:C₁₆H₃₀BaO₄
MDL Number:MFCD00058696