Silicon nitride α-phase ≥95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12033-89-5
:
Formula:N₄Si₃
MDL Number:MFCD00011230
:140.28 g/mol
Merck Index:14,8497
Melting Pt:1900 °C (subl.)
Density:3.2
Storage Temperature:Ambient
EINECS:234-796-8
Related Information
Frequently Bought Together