
2-Methyl-5-(trifluoromethoxy)benzylamine 96%
Catalog # H33791.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:205.18 g/mol
Storage Temperature:Ambient
UN:3267
ADR:8,III
:GHS05
CAS Number:1373920-87-6
Formula:C₉H₁₀F₃NO
MDL Number:MFCD22201056