4'-Benzyloxy-3'-methoxyacetophenone ≥98%
Catalog # L08430.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:1835-11-6
:
Formula:C₁₆H₁₆O₃
MDL Number:MFCD00017326
:256.30 g/mol
Melting Pt:85…87 °C
Storage Temperature:Ambient