Order Entry
Czech Republic
ContactUsLinkComponent
5'-Bromo-2'-hydroxy-3'-nitroacetophenone, Sigma-Aldrich®
5'-Bromo-2'-hydroxy-3'-nitroacetophenone, Sigma-Aldrich®
Catalog # SIAL534153-25G
Supplier:  Merck
CAS Number:  
5'-Bromo-2'-hydroxy-3'-nitroacetophenone, Sigma-Aldrich®
Catalog # SIAL534153-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :260.05 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:70978-54-0
Formula:C₈H₆BrNO₄
MDL Number:MFCD01631134