3-(N,N-Dimethylamino)phenylboronic acid, Sigma-Aldrich®
Catalog # SIAL680532-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:164.9974 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:178752-79-9
Formula:C₈H₁₂BNO₂
MDL Number:MFCD01318996