
4-(Ethoxycarbonyl)phenylboronic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4334-88-7
:
Formula:C₉H₁₁BO₄
MDL Number:MFCD02179441
:193.99 g/mol
Melting Pt:184…189 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Titration with NaOH:≥96.0 %
Specification Test Results
Frequently Bought Together