

Divinyl sulphone ≥97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:77-77-0
:
Formula:C₄H₆O₂S
MDL Number:MFCD00008623
:118.16 g/mol
Boiling Pt:233…234 °C
Melting Pt:∼–26 °C
Density:1.177
Flash Pt:102 °C (215 °F)
UN:2927
ADR:6.1,I
:Stabilised with hydroquinone (HQ)
EINECS:201-057-6
:GHS06
:GHS05
Storage Temperature:Refrigerator
Frequently Bought Together