1,1'-Azobis(N,N-dimethylformamide), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10465-78-8
:
Formula:C₆H₁₂N₄O₂
MDL Number:MFCD00008318
:172.19 g/mol
:M-Clarity™ quality level = MQ200
EINECS:233-951-7
Frequently Bought Together