

Acetone-[1,3-¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7217-25-6
:
Formula:C₃H₆O
MDL Number:MFCD00064448
ADR:3,II
:M-Clarity™ quality level = MQ200
UN:1090
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together