2,4,6-Trichloroanisole-[D5] 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA17714601AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:352439-08-8
- Description:2,4,6-Trichloroanisole-[D5] 100 µg/ml in acetonitrile
- Formula:C₇D₅Cl₃O
- Description:2,4,6-Trichloroanisole-[D5]