
Tetrabenzyl pyrophosphate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:990-91-0
:
Formula:C₂₈H₂₈O₇P₂
MDL Number:MFCD00051941
:538.48 g/mol
Melting Pt:58…64 °C
UN:3261
ADR:8,III
:GHS05
Related Information
Frequently Bought Together