Order Entry
Czech Republic
ContactUsLinkComponent
Scandium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade
Oxidizing
Irritant
Scandium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade
Catalog # 40579.NA
CAS Number:  
Oxidizing
Irritant
Scandium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade
Catalog # 40579.NA
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2 g
  • Seq:
    3107

Specifications

About this item

CAS Number:14066-05-8
 :
Formula:Cl₃O₁₂Sc
MDL Number:MFCD00243289
 :451.42 g/mol (343.30 anhy)
Storage Temperature:Ambient
EINECS:237-914-6
UN:3211
ADR:5.1,II
 :GHS03
 :GHS07