Order Entry
Czech Republic
ContactUsLinkComponent
Carbon disulphide-[¹³C] (99% ¹³C ), Sigma-Aldrich®
Flammable
Health
Irritant
Carbon disulphide-[¹³C] (99% ¹³C ), Sigma-Aldrich®
Catalog # SIAL486434-1G
Supplier:  Merck
CAS Number:  
Flammable
Health
Irritant
Carbon disulphide-[¹³C] (99% ¹³C ), Sigma-Aldrich®
Catalog # SIAL486434-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
ADR:3,I
 :M-Clarity™ quality level = MQ200
UN:1131
 :GHS02
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:30860-31-2
Formula:CS₂