Order Entry
Czech Republic
ContactUsLinkComponent
Acetophenone-[1,2-¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Acetophenone-[1,2-¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Catalog # SIAL487872-250MG
Supplier:  Merck
CAS Number:  
Acetophenone-[1,2-¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Catalog # SIAL487872-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
CAS Number:190314-15-9
Formula:C₈H₈O
MDL Number:MFCD00083868