
3-Chloro-2-methylphenylboronic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:313545-20-9
:
Formula:C₇H₈BClO₂
MDL Number:MFCD04115641
:170.40 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together