3-(1H-Pyrazol-1-yl)aniline
Catalog # APOSOR5682-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:159.1879 g/mol
CAS Number:184097-88-9
Formula:C₉H₉N₃
MDL Number:MFCD08059554