ContactUsLinkComponent
Cover spectacles, B-Line
Cover spectacles, B-Line
Catalog # 111-0343
Supplier:  Bollé Safety
undefined
Cover spectacles, B-Line
Catalog # 111-0343
Supplier:  Bollé Safety
Specifications
  • Type:
    Anti-UV (2.1.2), anti-scratch
  • Colour:
    Blue
  • Seq:
    02
  • Lenses:
    Clear PC
  • Pk:
    1
Specifications
About this item
Cover glasses (OTG) offering a panoramic field of vision.
  • Nylon frame
  • Scratch resistant
  • Wide side shields
Certifications: EN 166, (EN 170, EN 172)