Orders LinkContactUsLinkComponent
Powder triers
Powder triers
Supplier:   Sampling Systems
Powder triers
Supplier:  Sampling Systems
Certificates
Powder triers
3 Options Available Below
Product Details & Documents
These triers are suitable for taking a large volume, cross sectional sample from a container.
  • Robust all stainless steel construction with bright finish
  • Cutting scroll for the sampler to go into the product
  • Turn the handle to close the slot
Suitable for sampling cereals, grains, free flowing powders, granules and even slightly cohesive or compacted powders.
Certifications: GMP
Product Family Options
Product Information
  • Material
    Length
    No. of chambers
    Volume
    Availability
    Price
  • SAMP1216A-600
    Powder trier without bottle holder
    Powder trier without bottle holder
    Stainless steel 316
    600 mm
    3
    110 ml
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 42 830,00 Kč
    42 830,00 Kč
    1 * 1 KS
       42 830,00 Kč
  • SAMP1216A-1000
    Powder trier without bottle holder
    Powder trier without bottle holder
    Stainless steel 316L
    1000 mm
    4
    250 ml
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 49 960,00 Kč
    49 960,00 Kč
    1 * 1 KS
       49 960,00 Kč
  • SAMP1216A-1500
    Powder trier without bottle holder
    Powder trier without bottle holder
    Stainless steel 316
    1500 mm
    5
    300 ml
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 55 900,00 Kč
    55 900,00 Kč
    1 * 1 KS
       55 900,00 Kč