ContactUsLinkComponent
Pipette aid, Micro-Pipex
Pipette aid, Micro-Pipex
Catalog # HECH40558001
undefined
Pipette aid, Micro-Pipex
Catalog # HECH40558001
Specifications
  • Description:
    Pipetting aid, Micro-Pipex, for disposable micro pipettes, volume pipettes (RBC/WBC pipettes) and blood sugar pipettes up to 1 ml
  • Pk:
    1
Specifications
About this item
Aid for pipetting liquids by using disposable micropipettes with ring mark as well as other pipettes up to 1 ml, e.g. RBC and WBC pipettes.
  • Contaminated disposable micropipettes up to 50 µl are released contactless by the integrated ejection
  • Autoclavable at +121 °C
  • Pipette holder: max. 5 mm Ø
Please note: The pipetting aid may not be used for capillaries end-to-end.