Order Entry
Czech Republic
ContactUsLinkComponent
2,4-Dichloro-3-formylpyridine ≥97%
Irritant
2,4-Dichloro-3-formylpyridine ≥97%
Catalog # H34053.03
CAS Number:  
Irritant
2,4-Dichloro-3-formylpyridine ≥97%
Catalog # H34053.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :176.00 g/mol
Melting Pt:72…75 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:134031-24-6
Formula:C₆H₃Cl₂NO
MDL Number:MFCD07437909