
Acetoxyacetyl chloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13831-31-7
:
Formula:C₄H₅ClO₃
MDL Number:MFCD00011535
:136.53 g/mol
Boiling Pt:64…68 °C (25 mmHg)
Flash Pt:71 °C
Storage Temperature:Ambient
EINECS:237-542-4
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4255 to 1.4275 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together