


Thionyl chloride 1 M in dichloromethane
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7719-09-7
:
Formula:Cl₂OS
MDL Number:MFCD00011449
:118.97 g/mol
Density:1.35
Storage Temperature:Ambient
EINECS:231-748-8
UN:2922
ADR:8,III
:GHS05
:GHS08
:GHS07
Related Information
Frequently Bought Together