ContactUsLinkComponent
Silicon dioxide ≥99.995% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Silicon dioxide ≥99.995% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
  41099.KS
 :  
Silicon dioxide ≥99.995% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
  41099.KS
 :  
  • Pk:
    1
  • Seq:
    0001
 :7631-86-9
 :
 :SiO₂
 :MFCD00011232
 :60.08 g/mol
 :14,08493
 :2230 °C (1013 hPa)
 :1719 °C
 :2.2 g/cm³ (20 °C)
 :Ambient
 :231-545-4
 :14808-60-7