Order Entry
Czech Republic
ContactUsLinkComponent
Tris(triphenylphosphine)ruthenium(II) chloride ≥99%
Tris(triphenylphosphine)ruthenium(II) chloride ≥99%
Catalog # ABCRAB111898.0005
Supplier:  abcr
CAS Number:  
Tris(triphenylphosphine)ruthenium(II) chloride ≥99%
Catalog # ABCRAB111898.0005
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
 :958.85 g/mol
EINECS:239-569-7
CAS Number:15529-49-4
Formula:C₅₄H₄₅Cl₂P₃Ru
MDL Number:MFCD00013077