Order Entry
Czech Republic
ContactUsLinkComponent
(±)-1-(1-Naphthyl)ethylamine
(±)-1-(1-Naphthyl)ethylamine
Catalog # ABCRAB139259.0025
Supplier:  abcr
CAS Number:  
(±)-1-(1-Naphthyl)ethylamine
Catalog # ABCRAB139259.0025
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL

Specifications

About this item

 :
 :171.24 g/mol
CAS Number:42882-31-5
Formula:C₁₂H₁₃N
MDL Number:MFCD00004014