Order Entry
Czech Republic
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Palmitic Acid
  43051-100MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Palmitic Acid
  43051-100MG
 :  Merck
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    57-10-3
  • Description:
    Palmitic acid
  • Storage conditions:
    20...25 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00002747
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    1-Pentadecanecarboxylic acid; C16:0; Cetylic acid; Hexadecanoic acid; NSC 5030; PamOH
  • Melting point:
    61...62,5 °C (lit.)
  • Formula:
    CH₃(CH₂)₁₄COOH
  • Molecular weight:
    256,42
  • Density:
    0,852 g/ml at 25 °C (lit.)
  • Flash point:
    113 °C
  • Boiling point:
    271,5 °C/100 mm Hg (lit.)

 

 

 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.