Zinc standard solution, 1000 mg/L Zn in dil. nitric acid standard for ICP
Supplier: CPACHEM
Certificates
About this item
Frequently Bought Together
in Acetonitrile 1 * 1 mL
Supplier: EHRENSTORFER
Lyophilized Exosome Standard (U87 MG) 1 * 1 Vial
Supplier: Abcam
Tepraloxydim certified 1 * 0,1 g
Supplier: EHRENSTORFER
4'-Hydroxydiclofenac, Supelco®
Supplier: Merck
Beclomethasone Dipropionate, Reference Standards
Supplier: USP
Camphor 1 * 100 mg
Supplier: EHRENSTORFER
Supelco® TraceCERT® Organic Reference Standard, Propamocarb
Supplier: Merck
Fonofos certified 1 * 0,1 g
Supplier: EHRENSTORFER
Zinc standard solutions
Supplier: Hach
Calcium Standard for AAS, TraceCERT TM 1 * 250 mL
Supplier: Merck