Vanadium standard solution, 1000 mg/L V in dil. nitric acid standard for ICP
Supplier: CPACHEM
Certificates
About this item
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
pH buffer solutions
Supplier: Mettler - Toledo
pH buffer solutions, certified (DKD)
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
pH buffers, ready to use
Supplier: WTW
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond