Order Entry
Czech Republic
ContactUsLinkComponent
Phosphate standard solution, 1,000 mg/L PO₄³⁻ in Water standard for ion chromatography
Phosphate standard solution, 1,000 mg/L PO₄³⁻ in Water standard for ion chromatography
Catalog # ACSDH014.W.L25
Supplier:  CPACHEM
CAS Number:  
Phosphate standard solution, 1,000 mg/L PO₄³⁻ in Water standard for ion chromatography
Catalog # ACSDH014.W.L25
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    1000 mg/l
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    250 mL
  • Description:
    Phosphate standard solution

Specifications

About this item