
Palladium(II) chloride ≥99.9% (Pd basis)
Catalog # ABCRAB121380.0005
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
About this item
:
Merck Index:13,07058
Melting Pt:500 °C (dec.)
Density:4.0
Storage Temperature:Ambient
ADR:8,III
:177.33 g/mol
EINECS:231-596-2
UN:3260
:GHS07
CAS Number:7647-10-1
Formula:PdCl₂
MDL Number:MFCD00003558