Order Entry
Czech Republic
ContactUsLinkComponent
 
Herbicide reference standard, 3,6-Dichloropicolinic acid 100 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERXA11690000AL
 :  
Herbicide reference standard, 3,6-Dichloropicolinic acid 100 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERXA11690000AL
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    1702-17-6
  • Description:
    3,6-Dichloropicolinic acid 100 µg/ml in acetonitrile
  • Formula:
    C₆H₃Cl₂NO₂
  • Description:
    3,6-Dichloropicolinic acid